Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromopiperonal, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
15930-53-7
C8H5BrO3
229.03
CSQUXTSIDQURDV-UHFFFAOYSA-N
6-bromopiperonal,6-bromo-benzo 1,3 dioxole-5-carbaldehyde,2-bromo-4,5-methylenedioxybenzaldehyde,piperonal, 6-bromo,6-bromobenzo d 1,3 dioxole-5-carbaldehyde,6-bromo-1,3-benzodioxole-5-carboxaldehyde,6-bromo-2h-1,3-benzodioxole-5-carbaldehyde,1,3-benzodioxole-5-carboxaldehyde, 6-bromo,6-bromobenzo 1,3 dioxole-5-carbaldehyde,benzaldehyde, 2-bromo-4,5-methylenedioxy
95062
6-bromo-1,3-benzodioxole-5-carbaldehyde
C1OC2=C(O1)C=C(C(=C2)C=O)Br
15930-53-7
C8H5BrO3
229.03
CSQUXTSIDQURDV-UHFFFAOYSA-N
6-bromopiperonal,6-bromo-benzo 1,3 dioxole-5-carbaldehyde,2-bromo-4,5-methylenedioxybenzaldehyde,piperonal, 6-bromo,6-bromobenzo d 1,3 dioxole-5-carbaldehyde,6-bromo-1,3-benzodioxole-5-carboxaldehyde,6-bromo-2h-1,3-benzodioxole-5-carbaldehyde,1,3-benzodioxole-5-carboxaldehyde, 6-bromo,6-bromobenzo 1,3 dioxole-5-carbaldehyde,benzaldehyde, 2-bromo-4,5-methylenedioxy
95062
6-bromo-1,3-benzodioxole-5-carbaldehyde
C1OC2=C(O1)C=C(C(=C2)C=O)Br
Melting Point 125°C to 129°C
Color White to Yellow
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 229.03
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 6-Bromopiperonal
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning