Alfa Aesar™ 6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile, 98%

Manufacturer: Alfa Aesar™ H3370103

Quantity 1g

 View more versions of this product

Catalog No. AAH3370103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 352637-44-6
    Molecular Formula C6H3BrN4
    Formula Weight 223.03
    MDL Number MFCD00461392
    Synonym 6-Bromo-3-cyanopyrazolo[1,5-a]pyrimidine
    Chemical Name or Material 6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile

    Specifications

    Formula Weight 223.03
    Molecular Formula C6H3BrN4
    MDL Number MFCD00461392
    CAS 352637-44-6
    Melting Point 154° to 156°C
    Synonym 6-Bromo-3-cyanopyrazolo[1,5-a]pyrimidine
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 6-Bromopyrazolo[1,5-a]pyrimidine-3-carbonitrile
    Assay 98%

    GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit