Alfa Aesar™ 6-Bromoquinoline, 97%

Manufacturer: Alfa Aesar™ H55111MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55111MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5332-25-2
    Molecular Formula C9H6BrN
    Formula Weight 208.06
    MDL Number MFCD00024023
    Chemical Name or Material 6-Bromoquinoline

    Specifications

    Formula Weight 208.06
    TSCA TSCA
    Melting Point 24°C
    Quantity 250mg
    Flash Point >110°C (230°F)
    MDL Number MFCD00024023
    Molecular Formula C9H6BrN
    CAS 5332-25-2
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 6-Bromoquinoline
    Assay 97%

    Description

    Applications
    6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

    Solubility
    Soluble in acetone, acetonitrile, dichloromethane, ethyl acetate and THF.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit