Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Carboxyfluorescein, single isomer, 97%

Click to view available options
Quantity:
250mg
1g
3301-79-9
C21H12O7
376.32
MFCD00036873
BZTDTCNHAFUJOG-UHFFFAOYSA-N
6-carboxyfluorescein,6-fam,carboxyfluorescein,spiro isobenzofuran-1 3h ,9'-9h xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo,3,6,9-trihydroxyxanthen-9-yl terephthalic acid,3',6'-dihydroxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-6-carboxylic acid,3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid,spiro isobenzofuran-1 3h ,9'-9h xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo,5 6-carboxy fluorescein,bidd:gt0504
76806
CHEBI:39073
3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid
C1=CC2=C(C=C1C(=O)O)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O
3301-79-9
C21H12O7
376.32
MFCD00036873
BZTDTCNHAFUJOG-UHFFFAOYSA-N
6-carboxyfluorescein,6-fam,carboxyfluorescein,spiro isobenzofuran-1 3h ,9'-9h xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo,3,6,9-trihydroxyxanthen-9-yl terephthalic acid,3',6'-dihydroxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-6-carboxylic acid,3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid,spiro isobenzofuran-1 3h ,9'-9h xanthene-6-carboxylic acid, 3',6'-dihydroxy-3-oxo,5 6-carboxy fluorescein,bidd:gt0504
76806
CHEBI:39073
3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid
C1=CC2=C(C=C1C(=O)O)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O
Color Orange
Quantity 1g
Beilstein 54341
Formula Weight 376.32
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 6-Carboxyfluorescein, Single isomer

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

storageNote1 : 2° to 8°C