Learn More
6-Chloro-1-hexanol, 97%, Thermo Scientific Chemicals
Description
6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.
Solubility
Soluble in dimethylformamide.
Notes
Incompatible with strong oxidizing agents, strong bases, powdered metal salts, strong reducing agents, acid anhydrides and acid chlorides.
Specifications
Density | 1.027 |
Boiling Point | 109°C to 111°C (14 mmHg) |
Flash Point | 98°C (208°F) |
Refractive Index | 1.456 |
Quantity | 25 g |
Beilstein | 1697307 |
Solubility Information | Soluble in dimethylformamide. |
Formula Weight | 136.62 |
Percent Purity | 97% |
Chemical Name or Material | 6-Chloro-1-hexanol |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H315-H319 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 217-925-2 |
RTECSNumber | MO9160000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.