Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloro-1-hexene 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
928-89-2
C6H11Cl
118.604
MFCD00039391
BLMIXWDJHNJWDT-UHFFFAOYSA-N
6-chloro-1-hexene,1-hexene, 6-chloro,5-hexenyl chloride,6-chlorohexene,chloro-1-hex,6-chloro-1-hexen,1-chloro-5-hexene,1-chlorohex-5-ene,acmc-20alz4,5-hexen-1-yl chloride
70233
6-chlorohex-1-ene
C=CCCCCCl
928-89-2
C6H11Cl
118.604
MFCD00039391
BLMIXWDJHNJWDT-UHFFFAOYSA-N
6-chloro-1-hexene,1-hexene, 6-chloro,5-hexenyl chloride,6-chlorohexene,chloro-1-hex,6-chloro-1-hexen,1-chloro-5-hexene,1-chlorohex-5-ene,acmc-20alz4,5-hexen-1-yl chloride
70233
6-chlorohex-1-ene
C=CCCCCCl
Quantity 25g
Infrared Spectrum Authentic
Density 0.9070g/mL
Packaging Glass bottle
Color Colorless to Yellow
Refractive Index 1.4360 to 1.4380 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH2CH(CH2)4Cl
Formula Weight 118.61
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 6-Chloro-1-hexene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning