Alfa Aesar™ 6-Chloro-1-hexyne, 98%

Manufacturer: Alfa Aesar™ H53476AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH53476AA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10297-06-0
    Molecular Formula C6H9Cl
    Linear Formula Cl(CH2)4C≡CH
    Formula Weight 116.59
    MDL Number MFCD00013697
    UN Number UN1993
    Chemical Name or Material 6-Chloro-1-hexyne

    Specifications

    Formula Weight 116.59
    Linear Formula Cl(CH2)4C≡CH
    MDL Number MFCD00013697
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Boiling Point 83°C (95mmHg)
    Quantity 5mL
    Molecular Formula C6H9Cl
    CAS 10297-06-0
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 37°C (99°F)
    Chemical Name or Material 6-Chloro-1-hexyne
    Assay 98%

    Description

    Applications
    6-Chloro-1-hexyne is used as an organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit