Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloro-2-fluoro-3-methylbenzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
286474-59-7
C8H6ClFO
172.583
MFCD01631370
GGZOMBUJFMOEEZ-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methyl-benzaldehyde,6-chloro-2-fluoro-m-tolualdehyde,pubchem2895,acmc-1clwe,4-chloro-2-fluoro-3-formyltoluene,benzaldehyde, 6-chloro-2-fluoro-3-methyl,6-chloranyl-2-fluoranyl-3-methyl-benzaldehyde,6-chloro-2-fluoro-3-methylbenzaldehyde
1268278
6-chloro-2-fluoro-3-methylbenzaldehyde
CC1=C(C(=C(C=C1)Cl)C=O)F
286474-59-7
C8H6ClFO
172.583
MFCD01631370
GGZOMBUJFMOEEZ-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methyl-benzaldehyde,6-chloro-2-fluoro-m-tolualdehyde,pubchem2895,acmc-1clwe,4-chloro-2-fluoro-3-formyltoluene,benzaldehyde, 6-chloro-2-fluoro-3-methyl,6-chloranyl-2-fluoranyl-3-methyl-benzaldehyde,6-chloro-2-fluoro-3-methylbenzaldehyde
1268278
6-chloro-2-fluoro-3-methylbenzaldehyde
CC1=C(C(=C(C=C1)Cl)C=O)F
Quantity 5g
Flash Point >110°C (230°F)
Boiling Point 249°C
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning