Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloro-2-fluoro-3-methylbenzoyl chloride, 97%

Click to view available options
Quantity:
1g
5g
261762-81-6
C8H5Cl2FO
207.025
MFCD01631371
NWTPJKCARLIKFP-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methylbenzoylchloride,acmc-20alzl,2-fluoro-6-chloro-3-methyl-benzoyl chloride,6-chloro-2-fluoro-3-methyl-benzoyl chloride,benzoyl chloride,2-chloro-6-fluoro-3-methyl,6-chloranyl-2-fluoranyl-3-methyl-benzoyl chloride,benzoyl chloride, 6-chloro-2-fluoro-3-methyl-9ci
2773671
6-chloro-2-fluoro-3-methylbenzoyl chloride
CC1=C(C(=C(C=C1)Cl)C(=O)Cl)F
261762-81-6
C8H5Cl2FO
207.025
MFCD01631371
NWTPJKCARLIKFP-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methylbenzoylchloride,acmc-20alzl,2-fluoro-6-chloro-3-methyl-benzoyl chloride,6-chloro-2-fluoro-3-methyl-benzoyl chloride,benzoyl chloride,2-chloro-6-fluoro-3-methyl,6-chloranyl-2-fluoranyl-3-methyl-benzoyl chloride,benzoyl chloride, 6-chloro-2-fluoro-3-methyl-9ci
2773671
6-chloro-2-fluoro-3-methylbenzoyl chloride
CC1=C(C(=C(C=C1)Cl)C(=O)Cl)F
Quantity 1g
UN Number UN3265
Formula Weight 207.03
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.