Alfa Aesar™ 6-Chloro-2-fluoro-3-methylphenol, 97+%

Manufacturer: Alfa Aesar™ B2311306

Quantity 5g

 View more versions of this product

Catalog No. AAB2311306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 261762-91-8
    Molecular Formula C7H6ClFO
    Formula Weight 160.58
    MDL Number MFCD01631376
    UN Number UN3261
    Synonym 6-Chloro-2-fluoro-m-cresol
    Chemical Name or Material 6-Chloro-2-fluoro-3-methylphenol

    Specifications

    Formula Weight 160.58
    Molecular Formula C7H6ClFO
    UN Number UN3261
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 46° to 49°C
    MDL Number MFCD01631376
    CAS 261762-91-8
    Synonym 6-Chloro-2-fluoro-m-cresol
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Quantity 5g
    Chemical Name or Material 6-Chloro-2-fluoro-3-methylphenol
    Assay ≥97%

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit