Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloro-2-fluoronicotinic acid, 95%

Click to view available options
Quantity:
250mg
1g
1211578-46-9
C6H3ClFNO2
175.543
MFCD16610397
GSSVAHBRTISODE-UHFFFAOYSA-N
6-chloro-2-fluoronicotinic acid,6-chloro-2-fluoro nicotinic acid,6-chloro-2-fluoronicotinicacid,6-chloro-2-fluoropyridin-3-carboxylic acid,6-chloro-2-fluoro-pyridine-3-carboxylic acid
71464004
6-chloro-2-fluoropyridine-3-carboxylic acid
C1=CC(=NC(=C1C(=O)O)F)Cl
1211578-46-9
C6H3ClFNO2
175.543
MFCD16610397
GSSVAHBRTISODE-UHFFFAOYSA-N
6-chloro-2-fluoronicotinic acid,6-chloro-2-fluoro nicotinic acid,6-chloro-2-fluoronicotinicacid,6-chloro-2-fluoropyridin-3-carboxylic acid,6-chloro-2-fluoro-pyridine-3-carboxylic acid
71464004
6-chloro-2-fluoropyridine-3-carboxylic acid
C1=CC(=NC(=C1C(=O)O)F)Cl
Quantity 250mg
Formula Weight 175.54
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 6-Chloro-2-fluoronicotinic acid