Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-2-hexanone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
10226-30-9
C6H11ClO
134.603
MFCD00191625
CMDIDTNMHQUVPE-UHFFFAOYSA-N
6-chloro-2-hexanone,1-chloro-5-hexanone,2-hexanone, 6-chloro,1-chlorohexan-5-one,1-chlorohexane-5-one,4-chlorobutyl methyl ketone,chlorohexanone,1-chloro-hexan-5-one,6-chloro-hexan-2-one,6-chloranylhexan-2-one
82468
6-chlorohexan-2-one
CC(=O)CCCCCl
10226-30-9
C6H11ClO
134.603
MFCD00191625
CMDIDTNMHQUVPE-UHFFFAOYSA-N
6-chloro-2-hexanone,1-chloro-5-hexanone,2-hexanone, 6-chloro,1-chlorohexan-5-one,1-chlorohexane-5-one,4-chlorobutyl methyl ketone,chlorohexanone,1-chloro-hexan-5-one,6-chloro-hexan-2-one,6-chloranylhexan-2-one
82468
6-chlorohexan-2-one
CC(=O)CCCCCl
Packaging Glass bottle
Refractive Index 1.4425 to 1.4445
Quantity 25g
Color Yellow to Brown
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H3CCO(CH2)4Cl
Formula Weight 134.61
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 6-Chloro-2-hexanone, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 233-546-5

Recommended Storage : Product darkens during storage