Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloro-2-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
18368-63-3
C6H6ClN
127.57
MFCD00006245
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
6-chloro-2-picoline,6-chloro-2-methylpyridine,pyridine, 2-chloro-6-methyl,2-chloro-6-picoline,2-picoline, 6-chloro,unii-39mx1lq68l,2-chloro-6-methyl pyridine,2-chloro-6-methyl-pyridine,2-chloro-6-methy pyridine
87601
2-chloro-6-methylpyridine
CC1=NC(=CC=C1)Cl
18368-63-3
C6H6ClN
127.57
MFCD00006245
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
6-chloro-2-picoline,6-chloro-2-methylpyridine,pyridine, 2-chloro-6-methyl,2-chloro-6-picoline,2-picoline, 6-chloro,unii-39mx1lq68l,2-chloro-6-methyl pyridine,2-chloro-6-methyl-pyridine,2-chloro-6-methy pyridine
87601
2-chloro-6-methylpyridine
CC1=NC(=CC=C1)Cl
Flash Point 73°C
Boiling Point 64°C to 68°C (10.0mmHg)
Specific Gravity 1.16
Density 1.1600g/mL
Refractive Index 1.5260 to 1.528
Quantity 1g
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(C5H3N)Cl
Beilstein 20, I, 84
Formula Weight 127.57
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 6-chloro-2-picoline
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.