Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloro-3-(hydroxymethyl)pyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
21543-49-7
C6H6ClNO
143.57
GOXYBEXWMJZLJB-UHFFFAOYSA-N
2-chloro-5-hydroxymethylpyridine,6-chloropyridin-3-yl methanol,2-chloro-5-hydroxymethyl pyridine,2-chloro-5-pyridinemethanol,6-chloropyridine-3-methanol,6-chloro-3-pyridinemethanol,6-chloro-3-pyridinyl methanol,3-pyridinemethanol, 6-chloro,6-chloro-3-pyridyl methanol,6-chloro-3-pyridyl methan-1-ol
177161
(6-chloropyridin-3-yl)methanol
C1=CC(=NC=C1CO)Cl
21543-49-7
C6H6ClNO
143.57
GOXYBEXWMJZLJB-UHFFFAOYSA-N
2-chloro-5-hydroxymethylpyridine,6-chloropyridin-3-yl methanol,2-chloro-5-hydroxymethyl pyridine,2-chloro-5-pyridinemethanol,6-chloropyridine-3-methanol,6-chloro-3-pyridinemethanol,6-chloro-3-pyridinyl methanol,3-pyridinemethanol, 6-chloro,6-chloro-3-pyridyl methanol,6-chloro-3-pyridyl methan-1-ol
177161
(6-chloropyridin-3-yl)methanol
C1=CC(=NC=C1CO)Cl
Specific Gravity 1.28
Flash Point 109°C
Melting Point 46.7°C to 48.5°C
Color White to Brown
Quantity 5g
Boiling Point 190.0°C
Infrared Spectrum Authentic
Density 1.2800g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 143.57
Physical Form Powder or Low Melting Solid
Percent Purity 98%
Chemical Name or Material 6-chloro-3-(hydroxymethyl)pyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning