Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloro-3-nitro-2-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
22280-60-0
C6H5ClN2O2
172.5
MFCD00234160
GHSRMSJVYMITDX-UHFFFAOYSA-N
2-chloro-5-nitro-6-methylpyridine,6-chloro-3-nitro-2-picoline,2-chloro-5-nitro-6-picoline,6-chloro-2-methyl-3-nitro-pyridine,6-chloro-3-nitro-2-methylpyridine,pyridine, 6-chloro-2-methyl-3-nitro,2-chloro-6-methyl-5-nitropyridine,2-chloro-5-nitro-6-picoline 2-chloro-6-methyl-5-nitropyridine,pubchem6225,acmc-1cg7s
253123
6-chloro-2-methyl-3-nitropyridine
CC1=C(C=CC(=N1)Cl)[N+](=O)[O-]
22280-60-0
C6H5ClN2O2
172.5
MFCD00234160
GHSRMSJVYMITDX-UHFFFAOYSA-N
2-chloro-5-nitro-6-methylpyridine,6-chloro-3-nitro-2-picoline,2-chloro-5-nitro-6-picoline,6-chloro-2-methyl-3-nitro-pyridine,6-chloro-3-nitro-2-methylpyridine,pyridine, 6-chloro-2-methyl-3-nitro,2-chloro-6-methyl-5-nitropyridine,2-chloro-5-nitro-6-picoline 2-chloro-6-methyl-5-nitropyridine,pubchem6225,acmc-1cg7s
253123
6-chloro-2-methyl-3-nitropyridine
CC1=C(C=CC(=N1)Cl)[N+](=O)[O-]
Color Beige to Brown
Melting Point 54.0°C to 58.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 172.5
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 6-chloro-3-nitro-2-picoline
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning