Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline, 97+%

Click to view available options
Quantity:
1g
5g
18706-21-3
C10H5ClF3NO
247.601
MFCD00276583
MGEMMMJZAWYWNI-UHFFFAOYSA-N
6-chloro-4-hydroxy-2-trifluoromethyl quinoline,6-chloro-2-trifluoromethyl quinolin-4-ol,6-chloro-2-trifluoromethyl-1h-quinolin-4-one,6-chloro-2-trifluoromethyl-quinolin-4-ol,6-chloro-2-trifluoromethyl quinolin-4 1h-one,6-chloro-2-trifluoromethylquinolin-4-ol,acmc-1boyc,6-chloro-2-trifluoromethyl quinoline-4-ol,2-trifluoromethyl-4-hydroxy-6-chloroquinoline
2736609
6-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)C(=O)C=C(N2)C(F)(F)F
18706-21-3
C10H5ClF3NO
247.601
MFCD00276583
MGEMMMJZAWYWNI-UHFFFAOYSA-N
6-chloro-4-hydroxy-2-trifluoromethyl quinoline,6-chloro-2-trifluoromethyl quinolin-4-ol,6-chloro-2-trifluoromethyl-1h-quinolin-4-one,6-chloro-2-trifluoromethyl-quinolin-4-ol,6-chloro-2-trifluoromethyl quinolin-4 1h-one,6-chloro-2-trifluoromethylquinolin-4-ol,acmc-1boyc,6-chloro-2-trifluoromethyl quinoline-4-ol,2-trifluoromethyl-4-hydroxy-6-chloroquinoline
2736609
6-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
C1=CC2=C(C=C1Cl)C(=O)C=C(N2)C(F)(F)F
Melting Point 278°C to 280°C
Quantity 1g
Formula Weight 247.61
Percent Purity ≥97%
Chemical Name or Material 6-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures