Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-4-hydroxycoumarin, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
19484-57-2
C9H5ClO3
196.586
MFCD00239399
HUMZENGQNOATEQ-UHFFFAOYSA-N
6-chloro-4-hydroxycoumarin,6-chloro-4-hydroxy-2h-chromen-2-one,4-hydroxy-6-chlorocoumarin,6-chloro-4-hydroxy-2h-1-benzopyran-2-one,2h-1-benzopyran-2-one,6-chloro-4-hydroxy,6-chloro-2-hydroxychromen-4-one,6-chloro-2-hydroxy-4h-chromen-4-one,pubchem24343,acmc-1bsui,6-chlor-4-oxy-cumarin
54686348
6-chloro-4-hydroxychromen-2-one
C1=CC2=C(C=C1Cl)C(=CC(=O)O2)O
19484-57-2
C9H5ClO3
196.586
MFCD00239399
HUMZENGQNOATEQ-UHFFFAOYSA-N
6-chloro-4-hydroxycoumarin,6-chloro-4-hydroxy-2h-chromen-2-one,4-hydroxy-6-chlorocoumarin,6-chloro-4-hydroxy-2h-1-benzopyran-2-one,2h-1-benzopyran-2-one,6-chloro-4-hydroxy,6-chloro-2-hydroxychromen-4-one,6-chloro-2-hydroxy-4h-chromen-4-one,pubchem24343,acmc-1bsui,6-chlor-4-oxy-cumarin
54686348
6-chloro-4-hydroxychromen-2-one
C1=CC2=C(C=C1Cl)C(=CC(=O)O2)O
Packaging Glass bottle
Melting Point 270°C to 275°C
Color Beige
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 196.59
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 6-Chloro-4-hydroxycoumarin
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.