Learn More
eMolecules​ 6-chloro-4-iodo-1H-indazole | 885519-56-2 | 100G DFS Item
Manufacturer:  eMolecules​ 388203597
Pharmablock - Stock | 6-chloro-4-iodo-1H-indazole | 100G | 885519-56-2 | MFCD10698507 | MW:271.34
Catalog No. 50-160-9543
Due to product restrictions, please to purchase or view availability for this product.