Alfa Aesar™ 6-Chloro-5-fluorobenzimidazole, 95%

Manufacturer: Alfa Aesar™ A1090406

Quantity 5g

 View more versions of this product

Catalog No. AAA1090406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 175135-04-3
    Molecular Formula C7H4ClFN2
    Formula Weight 170.56
    MDL Number MFCD00156132
    Chemical Name or Material 6-Chloro-5-fluorobenzimidazole

    Specifications

    Formula Weight 170.56
    CAS 175135-04-3
    Molecular Formula C7H4ClFN2
    MDL Number MFCD00156132
    Melting Point 148° to 150°C
    Quantity 5g
    Chemical Name or Material 6-Chloro-5-fluorobenzimidazole
    Assay 95%

    Description

    Applications
    6-Chloro-5-fluorobenzimidazole is used as a pharamceutical intermediate, agrochemicals. As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment; subsequently several of the top pharmaceutical drugs contain fluorine.

    Solubility
    Insoluble in both water and organic solvents.

    Notes
    Stable under recommended storage conditions. Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit