Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloro-5-fluorobenzimidazole, 95%

Manufacturer:  Alfa Aesar™ A1090414

Quantity 25g

 View more versions of this product

Catalog No. AAA1090414


  Check Availability
Add to cart

Specifications

Description

Applications
6-Chloro-5-fluorobenzimidazole is used as a pharamceutical intermediate, agrochemicals. As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment; subsequently several of the top pharmaceutical drugs contain fluorine.

Solubility
Insoluble in both water and organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

175135-04-3
170.56
6-Chloro-5-fluorobenzimidazole
C7H4ClFN2
MFCD00156132

Specifications

148° to 150°C
175135-04-3
170.56
6-Chloro-5-fluorobenzimidazole
25g
C7H4ClFN2
MFCD00156132
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit