Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-5-nitro-2-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
56057-19-3
C6H5ClN2O2
172.5
MFCD03085820
UIEVSGOVFXWCIK-UHFFFAOYSA-N
6-chloro-5-nitro-2-picoline,2-chloro-3-nitro-6-methylpyridine,2-chloro-3-nitro-6-picoline,2-chloro-3-nitro-6-methyl pyridine,2-chloro-6-methyl-3-nitro-pyridine,pyridine, 2-chloro-6-methyl-3-nitro,2-chloro-6-methyl-3-nitropyridine?,pubchem6175,acmc-209lqq
1268230
2-chloro-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])Cl
56057-19-3
C6H5ClN2O2
172.5
MFCD03085820
UIEVSGOVFXWCIK-UHFFFAOYSA-N
6-chloro-5-nitro-2-picoline,2-chloro-3-nitro-6-methylpyridine,2-chloro-3-nitro-6-picoline,2-chloro-3-nitro-6-methyl pyridine,2-chloro-6-methyl-3-nitro-pyridine,pyridine, 2-chloro-6-methyl-3-nitro,2-chloro-6-methyl-3-nitropyridine?,pubchem6175,acmc-209lqq
1268230
2-chloro-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])Cl
Quantity 1g
Melting Point 70°C to 74°C
Color Brown
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 172.5
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 6-Chloro-5-nitro-2-picoline
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning