Alfa Aesar™ 6-Chloro-7-iodo-7-deazapurine, 96%

Manufacturer: Alfa Aesar™ H5112806

Quantity 5g

 View more versions of this product

Catalog No. AAH5112806

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 123148-78-7
    Molecular Formula C6H3ClIN3
    Formula Weight 279.47
    MDL Number MFCD09263258
    Synonym 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
    Chemical Name or Material 6-Chloro-7-iodo-7-deazapurine

    Specifications

    Formula Weight 279.47
    MDL Number MFCD09263258
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point ≈180°C
    Quantity 5g
    Molecular Formula C6H3ClIN3
    CAS 123148-78-7
    Synonym 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 6-Chloro-7-iodo-7-deazapurine
    Assay 96%

    Description

    Applications
    6-Chloro-7-iodo-7-deazapurine is used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Light sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit