Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloroindole-2-carboxylic acid, 96%

Click to view available options
Quantity:
1g
5g
16732-75-5
C9H6ClNO2
195.602
MFCD01863157
BKPSJOSKWKTWAG-UHFFFAOYSA-N
6-chloroindole-2-carboxylic acid,pubchem1671,acmc-209dwc,chembl23134,6-chloro-2-indolecarboxylic acid,6-chloro indole-2-carboxylic acid,1h-indole-2-carboxylic acid, 6-chloro,6-chloro-1h-indole-2-carboxylic acid; 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-2-indolecarboxylic acid
85584
6-chloro-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1Cl)NC(=C2)C(=O)O
16732-75-5
C9H6ClNO2
195.602
MFCD01863157
BKPSJOSKWKTWAG-UHFFFAOYSA-N
6-chloroindole-2-carboxylic acid,pubchem1671,acmc-209dwc,chembl23134,6-chloro-2-indolecarboxylic acid,6-chloro indole-2-carboxylic acid,1h-indole-2-carboxylic acid, 6-chloro,6-chloro-1h-indole-2-carboxylic acid; 6-chloro-1h-indole-2-carboxylic acid; 6-chloro-2-indolecarboxylic acid
85584
6-chloro-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1Cl)NC(=C2)C(=O)O
Quantity 1g
Beilstein 152660
Formula Weight 195.6
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 6-Chloroindole-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning