Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloronicotinamide, 98%

Click to view available options
Quantity:
1g
5g
25g
6271-78-9
C6H5ClN2O
156.569
MFCD00006242
ZIJAZUBWHAZHPL-UHFFFAOYSA-N
6-chloronicotinamide,3-pyridinecarboxamide, 6-chloro,nicotinamide, 6-chloro,6-chloro-3-pyridinecarboxamide,6-chloro-nicotinamide,acmc-1b4gd,6-chloronicotinic acid amide,3-carbamoyl-6-chloropyridine,ksc492o5t
80456
6-chloropyridine-3-carboxamide
C1=CC(=NC=C1C(=O)N)Cl
6271-78-9
C6H5ClN2O
156.569
MFCD00006242
ZIJAZUBWHAZHPL-UHFFFAOYSA-N
6-chloronicotinamide,3-pyridinecarboxamide, 6-chloro,nicotinamide, 6-chloro,6-chloro-3-pyridinecarboxamide,6-chloro-nicotinamide,acmc-1b4gd,6-chloronicotinic acid amide,3-carbamoyl-6-chloropyridine,ksc492o5t
80456
6-chloropyridine-3-carboxamide
C1=CC(=NC=C1C(=O)N)Cl
Melting Point 214°C to 216°C
Quantity 5g
Beilstein 116043
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 6-Chloronicotinamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning