Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloronicotinoyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
58757-38-3
C6H3Cl2NO
176
FMEBIWNKYZUWFV-UHFFFAOYSA-N
6-chloronicotinoyl chloride,6-chloronicotinoylchloride,2-chloropyridine-5-carbonyl chloride,3-pyridinecarbonylchloride, 6-chloro,3-pyridinecarbonyl chloride, 6-chloro,6-chloro-nicotinoyl chloride,pubchem18665,6-chloronicotinic chloride,acmc-1ay8e,6-chloro-nicotinic chloride
2782165
6-chloropyridine-3-carbonyl chloride
C1=CC(=NC=C1C(=O)Cl)Cl
58757-38-3
C6H3Cl2NO
176
FMEBIWNKYZUWFV-UHFFFAOYSA-N
6-chloronicotinoyl chloride,6-chloronicotinoylchloride,2-chloropyridine-5-carbonyl chloride,3-pyridinecarbonylchloride, 6-chloro,3-pyridinecarbonyl chloride, 6-chloro,6-chloro-nicotinoyl chloride,pubchem18665,6-chloronicotinic chloride,acmc-1ay8e,6-chloro-nicotinic chloride
2782165
6-chloropyridine-3-carbonyl chloride
C1=CC(=NC=C1C(=O)Cl)Cl
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 110°C
Melting Point 48.0°C to 50.0°C
Color White
Boiling Point 128.0°C to 130.0°C (23.0 Torr)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 176
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 6-chloronicotinoyl chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger