Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloropiperonyl Chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
25g
50g
Packaging:
Glass bottle
23468-31-7
C8H6Cl2O2
205.034
MFCD00005822
APJKOQPCHGXQBI-UHFFFAOYSA-N
6-chloropiperonyl chloride,5-chloro-6-chloromethyl-1,3-benzodioxole,1,3-benzodioxole, 5-chloro-6-chloromethyl,5-chloro-6-chloromethyl benzo d 1,3 dioxole,5-chloro-6-chloromethyl-2h-1,3-benzodioxole,6-chloro-5-chloromethyl-2h-benzo d 1,3-dioxolene,5-chloro-6-chloromethyl-benzo 1,3 dioxole,1-chloro-2-chloromethyl-4,5-methylenedioxybenzene,5-chloro-6-chloromethyl-1,3-benzodioxole; 2-chloro-4,5-methylenedioxy benzyl chloride
90113
5-chloro-6-(chloromethyl)-1,3-benzodioxole
C1OC2=C(O1)C=C(C(=C2)CCl)Cl
23468-31-7
C8H6Cl2O2
205.034
MFCD00005822
APJKOQPCHGXQBI-UHFFFAOYSA-N
6-chloropiperonyl chloride,5-chloro-6-chloromethyl-1,3-benzodioxole,1,3-benzodioxole, 5-chloro-6-chloromethyl,5-chloro-6-chloromethyl benzo d 1,3 dioxole,5-chloro-6-chloromethyl-2h-1,3-benzodioxole,6-chloro-5-chloromethyl-2h-benzo d 1,3-dioxolene,5-chloro-6-chloromethyl-benzo 1,3 dioxole,1-chloro-2-chloromethyl-4,5-methylenedioxybenzene,5-chloro-6-chloromethyl-1,3-benzodioxole; 2-chloro-4,5-methylenedioxy benzyl chloride
90113
5-chloro-6-(chloromethyl)-1,3-benzodioxole
C1OC2=C(O1)C=C(C(=C2)CCl)Cl
Melting Point 61°C to 65°C
Color White
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 205.04
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 6-Chloropiperonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger