Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Chloropyrazine-2-carboxylic acid, 95%

Click to view available options
Quantity:
250mg
1g
5g
23688-89-3
C5H3ClN2O2
158.541
MFCD00837871
KGGYMBKTQCLOTE-UHFFFAOYSA-N
6-chloro-pyrazine-2-carboxylic acid,6-chloro-2-pyrazinecarboxylic acid,2-carboxy-6-chloropyrazine,2-chloro-6-carboxypyrazine,pyrazinecarboxylic acid, 6-chloro,2-pyrazinecarboxylic acid, 6-chloro,6-chloropyrazine-2-carboxylicacid,6-chloro-pyrazine-2-carboxylicacid,pubchem16687,6-chloro-2-pyrazinoic acid
14500875
6-chloropyrazine-2-carboxylic acid
C1=C(N=C(C=N1)Cl)C(=O)O
23688-89-3
C5H3ClN2O2
158.541
MFCD00837871
KGGYMBKTQCLOTE-UHFFFAOYSA-N
6-chloro-pyrazine-2-carboxylic acid,6-chloro-2-pyrazinecarboxylic acid,2-carboxy-6-chloropyrazine,2-chloro-6-carboxypyrazine,pyrazinecarboxylic acid, 6-chloro,2-pyrazinecarboxylic acid, 6-chloro,6-chloropyrazine-2-carboxylicacid,6-chloro-pyrazine-2-carboxylicacid,pubchem16687,6-chloro-2-pyrazinoic acid
14500875
6-chloropyrazine-2-carboxylic acid
C1=C(N=C(C=N1)Cl)C(=O)O
Quantity 5g
Beilstein 879014
Formula Weight 158.54
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 6-Chloropyrazine-2-carboxylic acid

storageNote1 : -30° to -10°C