Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloropyridine-3-carbohydrazide, ≥95%, Maybridge

Click to view available options
Quantity:
10g
Packaging:
Amber Glass Bottle
168893-66-1
C6H6ClN3O
171.584
MFCD00221429
BIECNFYNFQQRRP-UHFFFAOYSA-N
6-chloronicotinohydrazide,2-chloropyridine-5-carbohydrazide,6-chloronicotinic acid hydrazide,pubchem18551,buttpark 9157-08,2-chloro-5-hydrazinocarbonyl pyridine,6-chloropyridine-3-carbohydrazide, 95+%,2-chloropyridine-5-carboxylic acid hydrazide,3-pyridinecarboxylicacid, 6-chloro-, hydrazide,3-pyridinecarboxylic acid, 6-chloro-, hydrazide
2800177
6-chloropyridine-3-carbohydrazide
C1=CC(=NC=C1C(=O)NN)Cl
168893-66-1
C6H6ClN3O
171.584
MFCD00221429
BIECNFYNFQQRRP-UHFFFAOYSA-N
6-chloronicotinohydrazide,2-chloropyridine-5-carbohydrazide,6-chloronicotinic acid hydrazide,pubchem18551,buttpark 9157-08,2-chloro-5-hydrazinocarbonyl pyridine,6-chloropyridine-3-carbohydrazide, 95+%,2-chloropyridine-5-carboxylic acid hydrazide,3-pyridinecarboxylicacid, 6-chloro-, hydrazide,3-pyridinecarboxylic acid, 6-chloro-, hydrazide
2800177
6-chloropyridine-3-carbohydrazide
C1=CC(=NC=C1C(=O)NN)Cl
Packaging Amber Glass Bottle
Quantity 10g
CAS Min % 95.0
Assay Percent Range 95%
Formula Weight 171.59
Percent Purity 95%
Chemical Name or Material 6-Chloropyridine-3-carbohydrazide

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.