Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chlorouracil, Acros Organics™

Click to view available options
Quantity:
1g
5g
4270-27-3
C4H3ClN2O2
146.53
PKUFNWPSFCOSLU-UHFFFAOYSA-N
6-chlorouracil,6-chloropyrimidine-2,4 1h,3h-dione,4-chlorouracil,6-chloropyrimidine-2,4-diol,6-chloro-2,4 1h,3h-pyrimidinedione,6-chloro-2,4-dihydroxypyrimidine,6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione,6-chloro-2,4-pyrimidinediol,6-chloro-1,3-dihydropyrimidine-2,4-dione,4-chloro-2,6-dihydroxypyrimidine
73272
6-chloro-1H-pyrimidine-2,4-dione
C1=C(NC(=O)NC1=O)Cl
4270-27-3
C4H3ClN2O2
146.53
PKUFNWPSFCOSLU-UHFFFAOYSA-N
6-chlorouracil,6-chloropyrimidine-2,4 1h,3h-dione,4-chlorouracil,6-chloropyrimidine-2,4-diol,6-chloro-2,4 1h,3h-pyrimidinedione,6-chloro-2,4-dihydroxypyrimidine,6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione,6-chloro-2,4-pyrimidinediol,6-chloro-1,3-dihydropyrimidine-2,4-dione,4-chloro-2,6-dihydroxypyrimidine
73272
6-chloro-1H-pyrimidine-2,4-dione
C1=C(NC(=O)NC1=O)Cl
NMR Consistent with structure (1H−NMR)
Infrared Spectrum Authentic
Quantity 5g
Formula Weight 146.53
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material 6-Chlorouracil

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

Warning