Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Cyano-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH59153MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624741-47-5
Molecular Formula C13H15BFNO2
Molecular Weight (g/mol) 247.076
MDL Number MFCD18729920
InChI Key HIMZNJDDVIROEC-UHFFFAOYSA-N
Synonym 3-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-fluoro-2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-fluoro-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 2-cyano-6-fluorophenyl boronic acid pinacol ester
PubChem CID 22240104
IUPAC Name 3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)C#N

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 247.08
Percent Purity 96%
Chemical Name or Material 6-Cyano-2-fluorobenzeneboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.