Learn More
Aat BioquestSupplier Diversity Partner 6-FAM [6-Carboxyfluorescein] *CAS 3301-79-9* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 106
6-FAM is the other purified isomer of 5(6)-carboxyfluorescein. Complimemtary to 5-FAM isomer, 6-FAM is mainly used in labeling nucleotides and nucleic acids.
Catalog No. 50-195-2716