Learn More
Aat BioquestSupplier Diversity Partner 6-FAM Alkyne DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 134
6-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group. It is widely used for labeling oligonucleotides. 6-FAM is the purified single isomer of carboxyfluorescein.
Catalog No. 50-195-2745