Learn More
Aat BioquestSupplier Diversity Partner 6-FAM Azide DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 955
6-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group. It is widely used for labeling oligonucleotides. 6-FAM is the purified single isomer of carboxyfluorescein.
Catalog No. 50-195-2744