Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoro-1-indanone, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1481-32-9
C9H7FO
150.15
LVUUCFIQQHEFEJ-UHFFFAOYSA-N
6-fluoro-1-indanone,6-fluoro-2,3-dihydro-1h-inden-1-one,6-fluoro-indan-1-one,6-fluoroindan-1-one,6-fluroro-1-indanone,1h-inden-1-one, 6-fluoro-2,3-dihydro,6-fluoroindanone,6fluoro-1-indanone,pubchem8856,acmc-1bwfy
1519464
6-fluoro-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=CC(=C2)F
1481-32-9
C9H7FO
150.15
LVUUCFIQQHEFEJ-UHFFFAOYSA-N
6-fluoro-1-indanone,6-fluoro-2,3-dihydro-1h-inden-1-one,6-fluoro-indan-1-one,6-fluoroindan-1-one,6-fluroro-1-indanone,1h-inden-1-one, 6-fluoro-2,3-dihydro,6-fluoroindanone,6fluoro-1-indanone,pubchem8856,acmc-1bwfy
1519464
6-fluoro-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=CC(=C2)F
Melting Point 58°C to 61°C
Packaging Glass Bottle
Quantity 1g
Color White to Brown
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 150.15
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 6-Fluoro-1-indanone
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning