Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoroindole, 98%, Thermo Scientific Chemicals

Catalog Number AAB2463314
Click to view available options
Quantity:
1 g
5 g
25 g
399-51-9
C8H6FN
135.14
MFCD00056933
YYFFEPUCAKVRJX-UHFFFAOYSA-N
6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g
351278
6-fluoro-1H-indole
FC1=CC=C2C=CNC2=C1
399-51-9
C8H6FN
135.14
MFCD00056933
YYFFEPUCAKVRJX-UHFFFAOYSA-N
6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g
351278
6-fluoro-1H-indole
FC1=CC=C2C=CNC2=C1

6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool place.Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 74°C to 77°C
Quantity 25 g
Beilstein 112192
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 135.14
Percent Purity 98%
Chemical Name or Material 6-Fluoroindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit