Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoroindole, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
399-51-9
C8H6FN
135.14
MFCD00056933
YYFFEPUCAKVRJX-UHFFFAOYSA-N
6-fluoroindole,1h-indole, 6-fluoro,6-fluoro indole,6-fluoroindol,6-fluoro-indole,pubchem1682,acmc-1cty0,ksc222e4t,6-fluoroindole 1g
351278
6-fluoro-1H-indole
C1=CC(=CC2=C1C=CN2)F
399-51-9
C8H6FN
135.14
MFCD00056933
YYFFEPUCAKVRJX-UHFFFAOYSA-N
6-fluoroindole,1h-indole, 6-fluoro,6-fluoro indole,6-fluoroindol,6-fluoro-indole,pubchem1682,acmc-1cty0,ksc222e4t,6-fluoroindole 1g
351278
6-fluoro-1H-indole
C1=CC(=CC2=C1C=CN2)F
Melting Point 72°C to 77°C
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Beige to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble in water
Formula Weight 135.14
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 6-Fluoroindole
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning