Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoropyridine-3-carboxaldehyde, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
677728-92-6
C6H4FNO
125.1
PZPNGWWKCSJKOS-UHFFFAOYSA-N
2-fluoropyridine-5-carbaldehyde,6-fluoronicotinaldehyde,2-fluoro-5-formylpyridine,2-fluoropyridine-5-carboxaldehyde,6-fluoro-3-pyridinecarboxaldehyde,3-pyridinecarboxaldehyde, 6-fluoro,6-fluoropyridine-3-carboxaldehyde,pubchem5150,zlchem 1162,acmc-1b2mv
16414246
6-fluoropyridine-3-carbaldehyde
C1=CC(=NC=C1C=O)F
677728-92-6
C6H4FNO
125.1
PZPNGWWKCSJKOS-UHFFFAOYSA-N
2-fluoropyridine-5-carbaldehyde,6-fluoronicotinaldehyde,2-fluoro-5-formylpyridine,2-fluoropyridine-5-carboxaldehyde,6-fluoro-3-pyridinecarboxaldehyde,3-pyridinecarboxaldehyde, 6-fluoro,6-fluoropyridine-3-carboxaldehyde,pubchem5150,zlchem 1162,acmc-1b2mv
16414246
6-fluoropyridine-3-carbaldehyde
C1=CC(=NC=C1C=O)F
Quantity 25g
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Formula Weight 125.1
Percent Purity 95%
Chemical Name or Material 6-Fluoropyridine-3-carboxaldehyde

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning