Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Fluoroquinoline-2-carboxaldehyde, 97%

Click to view available options
Quantity:
250mg
1g
260430-93-1
C10H6FNO
175.162
MFCD06751849
JDQMJKKGPXPPOJ-UHFFFAOYSA-N
2-quinolinecarboxaldehyde, 6-fluoro,6-fluoro-2-quinolinecarbaldehyde,6-fluoro-quinoline-2-carbaldehyde,6-fluoroquinoline-2-carboxaldehyde,4-aminomethyl-1-isoindolinone,6-fluoroquinoline-2-carboxaldehyde
3159605
6-fluoroquinoline-2-carbaldehyde
C1=CC2=C(C=CC(=N2)C=O)C=C1F
260430-93-1
C10H6FNO
175.162
MFCD06751849
JDQMJKKGPXPPOJ-UHFFFAOYSA-N
2-quinolinecarboxaldehyde, 6-fluoro,6-fluoro-2-quinolinecarbaldehyde,6-fluoro-quinoline-2-carbaldehyde,6-fluoroquinoline-2-carboxaldehyde,4-aminomethyl-1-isoindolinone,6-fluoroquinoline-2-carboxaldehyde
3159605
6-fluoroquinoline-2-carbaldehyde
C1=CC2=C(C=CC(=N2)C=O)C=C1F
Quantity 1g
Melting Point 113°C to 120°C
Sensitivity Moisture Sensitive
Beilstein 8479194
Formula Weight 175.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 6-Fluoroquinoline-2-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C