Learn More
Labconco™ 6 ft. Purifier Logic+ Class II Type A2 Biosafety Cabinet with 12 in. Sash Opening
SDP
6 ft. Purifier Logic+ Class II Type A2 Biosafety Cabinet with 12 in. Sash Opening creates a particulate-free environment for small animal research activities such as cage changing, weighing, anesthesia delivery and surgical procedures.
$20593.90 - $22502.80
Specifications
Description Class II Type A2 Biosafety Cabinet
Certifications/Compliance NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA
Chamber Material Stainless Steel
Model Logic+ A2 12
Outlets 2
View More Specs
Products 20
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
30-262-0021
SDP
View Documents
Labconco™
302620021
Japan 100 V, 50/60 Hz, 12 A Biosafety Cabinet Only
Each for $20,593.90
Add to Cart
30-262-0031
SDP
View Documents
Labconco™
302620031
Schuko 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $20,593.90
Add to Cart
30-262-0041
SDP
View Documents
Labconco™
302620041
China, Australia 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $20,593.90
Add to Cart
30-262-0051
SDP
View Documents
Labconco™
302620051
UK 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $20,593.90
Add to Cart
30-262-0071
SDP
View Documents
Labconco™
302620071
India 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $20,593.90
Add to Cart
30-262-0121
SDP
View Documents
Labconco™
302620121
Japan 100 V, 50/60 Hz, 12 A Base Stand
Each for $21,603.90
Add to Cart
30-262-0131
SDP
View Documents
Labconco™
302620131
Schuko 230 V, 50/60 Hz, 6 A Base Stand
Each for $21,603.90
Add to Cart
30-262-0141
SDP
View Documents
Labconco™
302620141
China, Australia 230 V, 50/60 Hz, 6 A Base Stand
Each for $21,603.90
Add to Cart
30-262-0151
SDP
View Documents
Labconco™
302620151
UK 230 V, 50/60 Hz, 6 A Base Stand
Each for $21,603.90
Add to Cart
30-262-0171
SDP
View Documents
Labconco™
302620171
India 230 V, 50/60 Hz, 6 A Base Stand
Each for $21,603.90
Add to Cart
30-262-1021
SDP
View Documents
Labconco™
302621021
Japan 100 V, 50/60 Hz, 12 A Accessory Pk.
Each for $21,492.80
Add to Cart
30-262-1031
SDP
View Documents
Labconco™
302621031
Schuko 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $21,492.80
Add to Cart
30-262-1041
SDP
View Documents
Labconco™
302621041
China, Australia 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $21,492.80
Add to Cart
30-262-1051
SDP
View Documents
Labconco™
302621051
UK 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $21,492.80
Add to Cart
30-262-1071
SDP
View Documents
Labconco™
302621071
India 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $21,492.80
Add to Cart
30-262-1121
SDP
View Documents
Labconco™
302621121
Japan 100 V, 50/60 Hz, 12 A Base Stand, Accessory Pk.
Each for $22,502.80
Add to Cart
30-262-1131
SDP
View Documents
Labconco™
302621131
Schuko 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $22,502.80
Add to Cart
30-262-1141
SDP
View Documents
Labconco™
302621141
China, Australia 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $22,502.80
Add to Cart
30-262-1151
SDP
View Documents
Labconco™
302621151
UK 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $22,502.80
Add to Cart
30-262-1171
SDP
View Documents
Labconco™
302621171
India 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $22,502.80
Add to Cart
Description
Description
  • Purifier Logic+ Class II A2 Biosafety Cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels
  • Designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection
  • Some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included
  • Inclination™ Technology Suite that includes MyLogic™ operating system with eye-level color display, 10° angled sash, patented curved air grille with built in armrest, plus fully accessible controls, outlets, and fixtures
  • MyLogic is easy to use, displaying the BSC's status with clear messaging and graphics that notify users of filter life, alerts, and alarms
  • Energy-efficient ECM blower powered by Labconco’s exclusive Constant Airflow Profile™ (CAP) technology
Specifications
Specifications
Class II Type A2 Biosafety Cabinet
Stainless Steel
2
78.3 in.
0.9999
198.9 cm
LED
5 Years
157.0 cm
81.3 cm
NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA
Logic+ A2 12
2
Full-color LCD
HEPA
Powder Coated Steel
12 in.
61.8 in.
32.7 in.
690 lb.