Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Hexadecanone, 97%

Click to view available options
Quantity:
5g
25g
57661-23-1
C16H32O
240.431
MFCD00051549
CIAHHJXQZXLMPN-UHFFFAOYSA-N
6-hexadecanone,acmc-20alyo
93752
hexadecan-6-one
CCCCCCCCCCC(=O)CCCCC
57661-23-1
C16H32O
240.431
MFCD00051549
CIAHHJXQZXLMPN-UHFFFAOYSA-N
6-hexadecanone,acmc-20alyo
93752
hexadecan-6-one
CCCCCCCCCCC(=O)CCCCC
Melting Point 34°C to 38°C
Quantity 25g
Formula Weight 240.43
Percent Purity 97%
Chemical Name or Material 6-Hexadecanone