Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hexanoyl-D-erythro-sphingosine, 99+%, synthetical, ACROS Organics™

Click to view available options
Quantity:
10mg
50mg
Packaging:
Glass bottle
124753-97-5
C24H47NO3
397.644
MFCD00171385
NPRJSFWNFTXXQC-PKTZIBPZSA-N
ceramide c6
71433696
N-[(2R,3S)-1,3-dihydroxyoctadec-4-en-2-yl]hexanamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)CCCCC)O
124753-97-5
C24H47NO3
397.644
MFCD00171385
NPRJSFWNFTXXQC-PKTZIBPZSA-N
ceramide c6
71433696
N-[(2R,3S)-1,3-dihydroxyoctadec-4-en-2-yl]hexanamide
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)CCCCC)O
Quantity 10mg
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 397.64
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material 6-Hexanoyl-D-erythro-sphingosine, 99+%
Show More Show Less