Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%

Click to view available options
Quantity:
250mg
1g
14446-24-3
C9H11NO
149.193
MFCD01717047
SCMZIFSYPJICCV-UHFFFAOYSA-N
6-hydroxy-1,2,3,4-tetrahydroisoquinoline,1,2,3,4-tetrahydro-isoquinolin-6-ol,1,2,3,4-tetrahydro-6-isoquinolinol,6-isoquinolinol, 1,2,3,4-tetrahydro,pubchem13941,6-hydroxytetrahydroisochinolin,ksc376k4j,6-hydroxy-1,2,3,4-tetrahydro-isoquinoline
26698
1,2,3,4-tetrahydroisoquinolin-6-ol
C1CNCC2=C1C=C(C=C2)O
14446-24-3
C9H11NO
149.193
MFCD01717047
SCMZIFSYPJICCV-UHFFFAOYSA-N
6-hydroxy-1,2,3,4-tetrahydroisoquinoline,1,2,3,4-tetrahydro-isoquinolin-6-ol,1,2,3,4-tetrahydro-6-isoquinolinol,6-isoquinolinol, 1,2,3,4-tetrahydro,pubchem13941,6-hydroxytetrahydroisochinolin,ksc376k4j,6-hydroxy-1,2,3,4-tetrahydro-isoquinoline
26698
1,2,3,4-tetrahydroisoquinolin-6-ol
C1CNCC2=C1C=C(C=C2)O
Melting Point 192°C to 193°C
Quantity 250mg
Beilstein 127578
Formula Weight 149.19
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-1,2,3,4-tetrahydroisoquinoline

RTECSNumber : NX5898000