Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-Hydroxy-1-tetralone 98.0+%, TCI America™
Click to view available options
Quantity:
5g
25g
Specifications
Color | Undesignated |
Melting Point | 155°C |
Quantity | 25g |
Formula Weight | 162.19 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥98.0% (GC,T) |
Chemical Name or Material | 6-Hydroxy-1-tetralone |
Safety and Handling
TSCA : No