Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Hydroxy-2-methylquinoline, 98%

Click to view available options
Quantity:
1g
5g
25g
613-21-8
C10H9NO
159.188
MFCD00047600
USSQQASIZNTRAJ-UHFFFAOYSA-N
6-hydroxy-2-methylquinoline,6-hydroxyquinaldine,2-methyl-6-quinolinol,6-quinolinol, 2-methyl,2-methyl-6-hydroxyquinoline,2-methyl-quinolin-6-ol,acmc-1bit8,2-methyl-6-quinolinol #,6-hydroxy-2-methyl quionoline,6-hydroxy-2-methylquinoline 1g
594395
2-methylquinolin-6-ol
CC1=NC2=C(C=C1)C=C(C=C2)O
613-21-8
C10H9NO
159.188
MFCD00047600
USSQQASIZNTRAJ-UHFFFAOYSA-N
6-hydroxy-2-methylquinoline,6-hydroxyquinaldine,2-methyl-6-quinolinol,6-quinolinol, 2-methyl,2-methyl-6-hydroxyquinoline,2-methyl-quinolin-6-ol,acmc-1bit8,2-methyl-6-quinolinol #,6-hydroxy-2-methyl quionoline,6-hydroxy-2-methylquinoline 1g
594395
2-methylquinolin-6-ol
CC1=NC2=C(C=C1)C=C(C=C2)O
Quantity 5g
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 6-Hydroxy-2-methylquinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No