Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Hydroxy-2-naphthoic acid, 99%

Click to view available options
Quantity:
1g
5g
25g
16712-64-4
C11H8O3
188.182
MFCD00060070
KAUQJMHLAFIZDU-UHFFFAOYSA-N
6-hydroxy-2-naphthoic acid,2-naphthalenecarboxylic acid, 6-hydroxy,6-hydroxy-2-naphthalenecarboxylic acid,6-carboxy-2-naphthol,2-hydroxynaphthalene-6-carboxylic acid,6-hydroxy-beta-naphthoic acid,unii-72xa858i6p,6-hydroxy-naphthalene-2-carboxylic acid,2-naphthoic acid, 6-hydroxy,2-hydroxy-6-naphthoic acid
85557
6-hydroxynaphthalene-2-carboxylic acid
C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O
16712-64-4
C11H8O3
188.182
MFCD00060070
KAUQJMHLAFIZDU-UHFFFAOYSA-N
6-hydroxy-2-naphthoic acid,2-naphthalenecarboxylic acid, 6-hydroxy,6-hydroxy-2-naphthalenecarboxylic acid,6-carboxy-2-naphthol,2-hydroxynaphthalene-6-carboxylic acid,6-hydroxy-beta-naphthoic acid,unii-72xa858i6p,6-hydroxy-naphthalene-2-carboxylic acid,2-naphthoic acid, 6-hydroxy,2-hydroxy-6-naphthoic acid
85557
6-hydroxynaphthalene-2-carboxylic acid
C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O
Melting Point 248°C to 250°C
Quantity 25g
Beilstein 2692084
Formula Weight 188.18
Percent Purity 99%
Chemical Name or Material 6-Hydroxy-2-naphthoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QN3722500

TSCA : TSCA