Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxy-2H-chromene-3-carboxaldehyde, 97%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
134822-76-7
C10H8O3
176.171
MFCD01076587
JSPGOHYLKVWWKI-UHFFFAOYSA-N
6-hydroxychromene-3-carboxaldehyde, acmc-20alym, 6-hydroxy-2h-3-chromenecarbaldehyde, 6-hydroxy-2h-1-benzopyran-3-carboxaldehyde, 2h-1-benzopyran-3-carboxaldehyde, 6-hydroxy
4770815
6-hydroxy-2H-chromene-3-carbaldehyde
C1C(=CC2=C(O1)C=CC(=C2)O)C=O
134822-76-7
C10H8O3
176.171
MFCD01076587
JSPGOHYLKVWWKI-UHFFFAOYSA-N
6-hydroxychromene-3-carboxaldehyde, acmc-20alym, 6-hydroxy-2h-3-chromenecarbaldehyde, 6-hydroxy-2h-1-benzopyran-3-carboxaldehyde, 2h-1-benzopyran-3-carboxaldehyde, 6-hydroxy
4770815
6-hydroxy-2H-chromene-3-carbaldehyde
C1C(=CC2=C(O1)C=CC(=C2)O)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 164°C to 166°C
Beilstein 6646155
Quantity 250mg
Sensitivity Air Sensitive
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-2H-chromene-3-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures