Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxy-3-nitro-2-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
28489-45-4
C6H6N2O3
154
MFCD00092010
AJDDHNXZBGZBJN-UHFFFAOYSA-N
6-methyl-5-nitropyridin-2-ol,2-hydroxy-6-methyl-5-nitropyridine,6-hydroxy-3-nitro-2-picoline,2-hydroxy-5-nitro-6-picoline,6-hydroxy-2-methyl-3-nitropyridine,6-methyl-5-nitropyridin-2 1h-one,2 1h-pyridinone, 6-methyl-5-nitro,6-methyl-5-nitro-2-pyridinol,6-methyl-5-nitro-pyridin-2-ol,pubchem10387
543053
6-methyl-5-nitro-1H-pyridin-2-one
CC1=C(C=CC(=O)N1)[N+](=O)[O-]
28489-45-4
C6H6N2O3
154
MFCD00092010
AJDDHNXZBGZBJN-UHFFFAOYSA-N
6-methyl-5-nitropyridin-2-ol,2-hydroxy-6-methyl-5-nitropyridine,6-hydroxy-3-nitro-2-picoline,2-hydroxy-5-nitro-6-picoline,6-hydroxy-2-methyl-3-nitropyridine,6-methyl-5-nitropyridin-2 1h-one,2 1h-pyridinone, 6-methyl-5-nitro,6-methyl-5-nitro-2-pyridinol,6-methyl-5-nitro-pyridin-2-ol,pubchem10387
543053
6-methyl-5-nitro-1H-pyridin-2-one
CC1=C(C=CC(=O)N1)[N+](=O)[O-]
Melting Point 230°C to 232°C
Color Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 154
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 6-Hydroxy-3-nitro-2-picoline
Show More Show Less

GHS P Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.