Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxycaproic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1191-25-9
C6H12O3
132.16
MFCD00046560
IWHLYPDWHHPVAA-UHFFFAOYSA-N
6-hydroxycaproic acid,6-hydroxyhexanoate,hexanoic acid, 6-hydroxy,6-hydroxy caproic acid,6-hydroxy-hexanoic acid,epsilon-hydroxycaproic acid,5-carboxypentanol,unii-3y3ox37nm8,6-hydroxycaproicacid,pract.,containslactone,6-hydroxy-caproic acid
14490
CHEBI:17869
6-hydroxyhexanoic acid
C(CCC(=O)O)CCO
1191-25-9
C6H12O3
132.16
MFCD00046560
IWHLYPDWHHPVAA-UHFFFAOYSA-N
6-hydroxycaproic acid,6-hydroxyhexanoate,hexanoic acid, 6-hydroxy,6-hydroxy caproic acid,6-hydroxy-hexanoic acid,epsilon-hydroxycaproic acid,5-carboxypentanol,unii-3y3ox37nm8,6-hydroxycaproicacid,pract.,containslactone,6-hydroxy-caproic acid
14490
CHEBI:17869
6-hydroxyhexanoic acid
C(CCC(=O)O)CCO
Specific Gravity 0.981
Melting Point 38.0°C to 40.0°C
Boiling Point 113.0°C to 116.0°C
Quantity 1g
Density 0.9810g/mL
Packaging Glass bottle
Water may contain variable amounts of water
Infrared Spectrum Authentic
Color White
Refractive Index 1.44
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula OH(CH2)5COOH
Formula Weight 132.16
Physical Form Low Melting Crystalline Solid
Percent Purity 95%
Chemical Name or Material 6-Hydroxycaproic acid, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning