Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxydopamine Hydrobromide 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
636-00-0
C8H12BrNO3
250.092
MLACDGUOKDOLGC-UHFFFAOYSA-N
6-hydroxydopamine hydrobromide,oxidopamine hydrobromide,6-hydroxydopamine hydrogen bromide,unii-twc1d4w0wb,oxidopamine hydrobromide,6-hydroxydopamine hbr,5-2-aminoethyl benzene-1,2,4-triol hydrobromide,twc1d4w0wb,hydroxydopamine hydrobromide,6-oh-da
176170
5-(2-aminoethyl)benzene-1,2,4-triol;hydrobromide
C1=C(C(=CC(=C1O)O)O)CCN.Br
636-00-0
C8H12BrNO3
250.092
MLACDGUOKDOLGC-UHFFFAOYSA-N
6-hydroxydopamine hydrobromide,oxidopamine hydrobromide,6-hydroxydopamine hydrogen bromide,unii-twc1d4w0wb,oxidopamine hydrobromide,6-hydroxydopamine hbr,5-2-aminoethyl benzene-1,2,4-triol hydrobromide,twc1d4w0wb,hydroxydopamine hydrobromide,6-oh-da
176170
5-(2-aminoethyl)benzene-1,2,4-triol;hydrobromide
C1=C(C(=CC(=C1O)O)O)CCN.Br
Color Beige to Brown
Quantity 250mg
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Linear Formula (HO)3C6H2CH2CH2NH2.HBr
Formula Weight 250.1
Physical Form Fine Crystalline Powder
Percent Purity 98.5 to 101.5% (total base), ≥98.5% (TLC)
Chemical Name or Material 6-Hydroxydopamine hydrobromide, 99%

EINECSNumber : 211-247-0

RTECSNumber : DC4600000