Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-Hydroxyindole, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Color | White |
Melting Point | 124.0°C to 130.0°C |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Quantity | 5g |
Assay Percent Range | 97.5% min. (HPLC) |
Formula Weight | 133.15 |
Physical Form | Shiny Crystalline Powder |
Percent Purity | 97% |
Chemical Name or Material | 6-Hydroxyindole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti
GHS Signal Word: Danger
Recommended Storage : Product may darken during storage